missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl Tetrazole, TRC

Product Code. 30477584
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30477584

Brand: Toronto Research Chemicals TRCM33025550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methyl Tetrazole
CAS 4076-36-2
Molecular Formula C2H4N4
Quantity 50 g
Synonym 5-Methyl-1,2,3,4-tetrazole, 5-Methyl-1H-tetrazole, 5-Methyl-2H-tetrazole, 5-Methyltetrazole, NSC 11136, T 2588F
InChI Formula InChI=1S/C2H4N4/c1-2-3-5-6-4-2/h1H3,(H,3,4,5,6)
SMILES Cc1nnn[nH]1
IUPAC Name 5-methyl-1H-tetrazole
Molecular Weight (g/mol) 84.08
Formula Weight 84.0436
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.