accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOCl

CAS RN 40877-09-6

IUPAC Name: 4-chloro-1-(4-chlorophenyl)butan-1-one
Synonyms: 4-chloro-1-(4-chlorophenyl)butan-1-one
Molecular Weight (g/mol): 217.09
Molecular Formula: C10H10Cl2O
InChi Key: UKCHLVFIVJBCKE-UHFFFAOYSA-N
SMILES: ClCCCC(=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)