accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ON

CAS RN 4113-04-6

IUPAC Name: quinoline-6-carbaldehyde
Synonyms: quinoline-6-carbaldehyde
Molecular Weight (g/mol): 157.17
Molecular Formula: C10H7NO
InChi Key: VUAOIXANWIFYCU-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2N=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results