accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OH3NOCH3

CAS RN 41365-75-7

IUPAC Name: 3,3-diethoxypropan-1-aminium
Synonyms: 3,3-diethoxypropan-1-aminium
Molecular Weight (g/mol): 148.23
Molecular Formula: C7H18NO2
InChi Key: PXXMSHBZYAOHBD-UHFFFAOYSA-O
SMILES: CCOC(CC[NH3+])OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results