CAS RN 41394-05-2
IUPAC Name:
4-amino-3-methyl-6-phenyl-4,5-dihydro-1,2,4-triazin-5-one
Synonyms:
metamitron methiamitron metamiton herbrak goltix metamitrone
Molecular Weight (g/mol):
202.22
Molecular Formula:
C10H10N4O
InChi Key:
VHCNQEUWZYOAEV-UHFFFAOYSA-N
SMILES:
CC1=NN=C(C(=O)N1N)C1=CC=CC=C1
1
–
5
of
5
results
2
Metamitron, TRC
CAS: 41394-05-2 Molecular Formula: C10 H10 N4 O Molecular Weight (g/mol): 202.21 Synonym: 4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5(4H)-one,3-Methyl-4-amino-6-phenyl-1,2,4-triazin-5-one,4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5-one,BAY 6676,BAY-DRW 1139,Burakomitron,DRW 1139,Gladiator,Goltix,Goltix Compact,Goltix Top,Goltix VM 90,Goltix WG,Goltix WG 70,Metafol 700SC,Metamitron,Metron 700SL,Metron Stefes IUPAC Name: 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one SMILES: CC1=NN=C(C(=O)N1N)c2ccccc2