missing translation for 'onlineSavingsMsg'
Learn More

Metamitron, TRC

Product Code. 30011060
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011060 1 g 1g
30003263 5 g 5g
30003225 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011060 Supplier Toronto Research Chemicals Supplier No. TRCM2588431G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41394-05-2
Molecular Formula C10 H10 N4 O
Molecular Weight (g/mol) 202.21
Synonym 4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5(4H)-one, 3-Methyl-4-amino-6-phenyl-1,2,4-triazin-5-one, 4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5-one, BAY 6676, BAY-DRW 1139, Burakomitron, DRW 1139, Gladiator, Goltix, Goltix Compact, Goltix Top, Goltix VM 90, Goltix WG, Goltix WG 70, Metafol 700SC, Metamitron, Metron 700SL, Metron Stefes
IUPAC Name 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one
SMILES CC1=NN=C(C(=O)N1N)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H10N4O/c1-7-12-13-9(10(15)14(7)11)8-5-3-2-4-6-8/h2-6H,11H2,1H3
Formula Weight 202.0855
Chemical Name or Material Metamitron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.