accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONF

CAS RN 41538-36-7

IUPAC Name: 4-(4-fluorobenzoyl)pyridine
Synonyms: 4-(4-fluorobenzoyl)pyridine
Molecular Weight (g/mol): 201.2
Molecular Formula: C12H8FNO
InChi Key: WTRWBYGUMQEFFI-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results