accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 4160-51-4

IUPAC Name: 1-(4-methoxyphenyl)butan-1-one
Synonyms: 1-(4-methoxyphenyl)butan-1-one
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: JLCDSZXBELPBRD-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=C(OC)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results