accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4160-63-8

IUPAC Name: (4-methoxyphenyl)(thiophen-2-yl)methanone
Synonyms: (4-methoxyphenyl)(thiophen-2-yl)methanone
Molecular Weight (g/mol): 218.27
Molecular Formula: C12H10O2S
InChi Key: KYVBFEMQEUXVQB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results