accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FBr

CAS RN 41604-19-7

IUPAC Name: 4-bromo-2-fluoro-1,1'-biphenyl
Synonyms: 4-bromo-2-fluoro-1,1'-biphenyl
Molecular Weight (g/mol): 251.1
Molecular Formula: C12H8BrF
InChi Key: HTRNHWBOBYFTQF-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)