accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Br2H2H2H2H2H

CAS RN 4165-57-5

IUPAC Name: 1-bromo(²H₅)benzene
Synonyms: 1-bromo(²H₅)benzene
Molecular Weight (g/mol): 162.04
Molecular Formula: C6H5Br
InChi Key: QARVLSVVCXYDNA-RALIUCGRSA-N
SMILES: [2H]C1=C([2H])C([2H])=C(Br)C([2H])=C1[2H]
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)