CAS RN 4165-96-2
IUPAC Name:
3-phenylpentanedioate
Synonyms:
3-phenylpentanedioate
Molecular Weight (g/mol):
206.2
Molecular Formula:
C11H10O4
InChi Key:
RZOKZOYSUCSPDF-UHFFFAOYSA-L
SMILES:
[O-]C(=O)CC(CC([O-])=O)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Phenylglutaric Acid, TRC
CAS: 4165-96-2 Molecular Formula: C11H12O4 Molecular Weight (g/mol): 208.21 Synonym: beta-Phenylglutaric Acid,3-Phenylpentanedioic Acid IUPAC Name: 3-phenylpentanedioic acid SMILES: OC(=O)CC(CC(=O)O)c1ccccc1