missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylglutaric Acid, TRC

Product Code. 30424391
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424391 100 mg -
30418085 250 mg 250mg
30418429 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424391 Supplier Toronto Research Chemicals Supplier No. TRCP327505100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-96-2
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.21
Synonym beta-Phenylglutaric Acid, 3-Phenylpentanedioic Acid
IUPAC Name 3-phenylpentanedioic acid
SMILES OC(=O)CC(CC(=O)O)c1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H12O4/c12-10(13)6-9(7-11(14)15)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,13)(H,14,15)
Formula Weight 208.0736
Chemical Name or Material 3-Phenylglutaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.