accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 41918-08-5

H3COO(S)CH3OCH3

CAS RN 41918-08-5

IUPAC Name: ethyl (2S)-2-methoxypropanoate
Synonyms: ethyl (2S)-2-methoxypropanoate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: WHRLOJCOIKOQGL-YFKPBYRVSA-N
SMILES: CCOC(=O)[C@H](C)OC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)