accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNO

CAS RN 420137-14-0

IUPAC Name: 3-(benzyloxy)pyrrolidine
Synonyms: 3-(benzyloxy)pyrrolidine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: CWBMYKUPMLRKQK-UHFFFAOYNA-N
SMILES: C(OC1CCNC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results