CAS RN 42019-07-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Fenofibric Acid Methyl Ester, TRC
CAS: 42019-07-8 Molecular Formula: C18 H17 Cl O4 Molecular Weight (g/mol): 332.78 Synonym: Methyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate,Fenofibrate Imp. D (EP) IUPAC Name: methyl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate SMILES: COC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2