accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHOHNO

CAS RN 4212-49-1

IUPAC Name: 5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-ethyl-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: RHIULBJJKFDJPR-UHFFFAOYSA-N
SMILES: CCC1=CNC(=O)NC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results