Learn More
5-Ethyluracil, 98+%, Thermo Scientific Chemicals
Product Code. 11313046
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313046 250 mg 250mg
11303046 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11313046 Supplier Thermo Scientific Alfa Aesar Supplier No. L10861.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
missing translation for 'validMainframeMsg'
Denna artikel kan inte returneras. Se returpolicy
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4212-49-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00079187
InChI Key RHIULBJJKFDJPR-UHFFFAOYSA-N
Synonym 5-ethyluracil, 5-ethyl uracil, 2,4-dihydroxy-5-ethylpyrimidine, 5-ethylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 5-ethyl, 5-ethyl-1,3-dihydropyrimidine-2,4-dione, 5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, uracil, 5-ethyl, zlchem 329, acmc-209jno
PubChem CID 73267
IUPAC Name 5-ethyl-1H-pyrimidine-2,4-dione
SMILES CCC1=CNC(=O)NC1=O
Specifikationer
Melting Point 308°C to 310°C
Quantity 250 mg
Beilstein 119494
Formula Weight 140.14
Percent Purity ≥98%
Chemical Name or Material 5-Ethyluracil
RUO – Research Use Only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.