accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HOHO

CAS RN 4254-15-3

IUPAC Name: propane-1,2-diol
Synonyms: 1,2-propanediol methylethylene glycol isopropylene glycol monopropylene glycol methylethyl glycol 1,2-propylenglykol α-propyleneglycol methyl glycol propylene glycol
Molecular Weight (g/mol): 76.1
Molecular Formula: C3H8O2
InChi Key: DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES: CC(O)CO
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)