accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4261-67-0

HClCH3ClNCH3CH3

CAS RN 4261-67-0

IUPAC Name: hydrogen (3-chloro-2-methylpropyl)dimethylamine chloride
Synonyms: hydrogen (3-chloro-2-methylpropyl)dimethylamine chloride
Molecular Weight (g/mol): 172.09
Molecular Formula: C6H15Cl2N
InChi Key: SOMIBONUMGNAEP-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(CCl)CN(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)