accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(R)(S)OO

CAS RN 42711-75-1

IUPAC Name: (5R,7S)-3-hydroxyadamantane-1-carboxylate
Synonyms: (5R,7S)-3-hydroxyadamantane-1-carboxylate
Molecular Weight (g/mol): 195.24
Molecular Formula: C11H15O3
InChi Key: CJJMAWPEZKYJAP-JZVMUCMXSA-M
SMILES: OC12C[C@@H]3C[C@H](C1)CC(C3)(C2)C([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)