accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4286-55-9

HOBr

CAS RN 4286-55-9

IUPAC Name: 6-bromohexan-1-ol
Synonyms: 6-bromo-1-hexanol C6H13BrO
Molecular Weight (g/mol): 181.07
Molecular Formula: C6H13BrO
InChi Key: FCMCSZXRVWDVAW-UHFFFAOYSA-N
SMILES: OCCCCCCBr
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)