accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OF

CAS RN 430-51-3

IUPAC Name: 1-fluoropropan-2-one
Synonyms: fluoroacetone
Molecular Weight (g/mol): 76.07
Molecular Formula: C3H5FO
InChi Key: MSWVMWGCNZQPIA-UHFFFAOYSA-N
SMILES: CC(=O)CF

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results