accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NON

CAS RN 43036-05-1

IUPAC Name: 3-(2-formyl-1H-pyrrol-1-yl)propanenitrile
Synonyms: 3-(2-formylpyrrol-1-yl)propanenitrile
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: SUUQLYBKCBUSAR-UHFFFAOYSA-N
SMILES: O=CC1=CC=CN1CCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results