CAS RN 43039-98-1
CAS RN 43039-98-1
IUPAC Name:
1-(1,3-thiazol-2-yl)propan-1-one
Synonyms:
thiazole, 2-propionyl-
Molecular Weight (g/mol):
141.19
Molecular Formula:
C6H7NOS
InChi Key:
TYRAENAWSLPSLW-UHFFFAOYSA-N
SMILES:
CCC(=O)C1=NC=CS1
1
–
1
of
1
results
1
2-Propionylthiazole, 99%
CAS: 43039-98-1 Molecular Formula: C6H7NOS Molecular Weight (g/mol): 141.188 MDL Number: MFCD01681404 InChI Key: TYRAENAWSLPSLW-UHFFFAOYSA-N Synonym: 2-propionylthiazole,1-thiazol-2-yl propan-1-one,2-propionyl thiazole,thiazole, 2-propionyl,1-propanone, 1-2-thiazolyl,1-2-thiazolyl-1-propanone,ethyl 2-thiazolyl ketone,1-1,3-thiazol-2-yl propan-1-one,2-propionylthiazol,unii-05cb62uh9k PubChem CID: 65288 IUPAC Name: 1-(1,3-thiazol-2-yl)propan-1-one SMILES: CCC(=O)C1=NC=CS1