accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClON

CAS RN 43111-31-5

IUPAC Name: 2-(2-chlorophenoxy)acetonitrile
Synonyms: 2-(2-chlorophenoxy)acetonitrile
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: JWRJGSQLKHGQOT-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1OCC#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)