accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 433926-48-8

OFFFClO

CAS RN 433926-48-8

IUPAC Name: 3-chloro-5-(trifluoromethoxy)benzaldehyde
Synonyms: 3-chloro-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: LWQSWEYYGGFLDR-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(Cl)=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results