accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 434-13-9

IUPAC Name: (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11aR)-7-hydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms: lithocholic acid 3α-hydroxy-5β-cholanoic acid
Molecular Weight (g/mol): 376.58
Molecular Formula: C24H40O3
InChi Key: SMEROWZSTRWXGI-HVATVPOCSA-N
SMILES: C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results