accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 434-64-0

IUPAC Name: 1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
Synonyms: perfluorotoluene
Molecular Weight (g/mol): 236.06
Molecular Formula: C7F8
InChi Key: USPWUOFNOTUBAD-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results