missing translation for 'onlineSavingsMsg'
Learn More

Octafluorotoluene, TRC

Product Code. 30423858
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423858 100 mg 100mg
30417124 50 mg 50mg
30418546 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423858 Supplier Toronto Research Chemicals Supplier No. TRCO273130100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-64-0
Molecular Formula C7F8
Molecular Weight (g/mol) 236.06
IUPAC Name 1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
SMILES Fc1c(F)c(F)c(c(F)c1F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7F8/c8-2-1(7(13,14)15)3(9)5(11)6(12)4(2)10
Formula Weight 235.9872
Chemical Name or Material Octafluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.