CAS RN 438450-43-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[3-Bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazol-5-yl]-6-chloro-3,8-dimethyl-4(3H)-quinazolinone, TRC
CAS: 438450-43-2 Molecular Formula: C18H12BrCl2N5O Molecular Weight (g/mol): 465.13 Synonym: Chlorantraniliprole Metabolite IN-EQW78,2-[3-Bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazol-5-yl]-6-chloro-3,8-dimethyl-4(3H)-quinazolinone (ACI),4(3H)-Quinazolinone, 2-[3-bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazol-5-yl]-6-chloro-3,8-dimethyl- (9CI, ACI) IUPAC Name: 2-[5-bromo-2-(3-chloropyridin-2-yl)pyrazol-3-yl]-6-chloro-3,8-dimethylquinazolin-4-one SMILES: CN1C(=O)c2cc(Cl)cc(C)c2N=C1c3cc(Br)nn3c4ncccc4Cl