missing translation for 'onlineSavingsMsg'
Learn More

2-[3-Bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazol-5-yl]-6-chloro-3,8-dimethyl-4(3H)-quinazolinone, TRC

Product Code. 30136038
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136038 50 mg 50mg
30134026 100 mg 100mg
30011928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136038 Supplier Toronto Research Chemicals Supplier No. TRCB44300050MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 438450-43-2
Molecular Formula C18H12BrCl2N5O
Molecular Weight (g/mol) 465.13
Synonym Chlorantraniliprole Metabolite IN-EQW78, 2-[3-Bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazol-5-yl]-6-chloro-3,8-dimethyl-4(3H)-quinazolinone (ACI), 4(3H)-Quinazolinone, 2-[3-bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazol-5-yl]-6-chloro-3,8-dimethyl- (9CI, ACI)
IUPAC Name 2-[5-bromo-2-(3-chloropyridin-2-yl)pyrazol-3-yl]-6-chloro-3,8-dimethylquinazolin-4-one
SMILES CN1C(=O)c2cc(Cl)cc(C)c2N=C1c3cc(Br)nn3c4ncccc4Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H12BrCl2N5O/c1-9-6-10(20)7-11-15(9)23-17(25(2)18(11)27)13-8-14(19)24-26(13)16-12(21)4-3-5-22-16/h3-8H,1-2H3
Formula Weight 462.9602
Percent Purity >95%
Chemical Name or Material 2-[3-Bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazol-5-yl]-6-chloro-3,8-dimethyl-4(3H)-quinazolinone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.