accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 446-51-5

HOF

CAS RN 446-51-5

IUPAC Name: (2-fluorophenyl)methanol
Synonyms: benzenemethanol, 2-fluoro-
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: QEHXDOJPVIHUDO-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)