missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl Alcohol, TRC

Product Code. 30404486
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404486 250 mg 250mg
30402106 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30404486 Supplier Toronto Research Chemicals Supplier No. TRCF598020250MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluorobenzyl Alcohol
CAS 446-51-5
Molecular Formula C7H7OF
Quantity 250 mg
Synonym NSC 158274, o-Fluorobenzyl Alcohol, 2-Fluorobenzenemethanol, (2-Fluorophenyl)methanol
InChI Formula InChI=1S/C7H7FO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5H2
SMILES C1=CC=C(C(=C1)CO)F
IUPAC Name (2-fluorophenyl)methanol
Molecular Weight (g/mol) 126.13
Formula Weight 126.0481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.