accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4466-24-4

H3CO

CAS RN 4466-24-4

IUPAC Name: 2-butylfuran
Synonyms: 2-butylfuran
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: NWZIYQNUCXUJJJ-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CO1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)