missing translation for 'onlineSavingsMsg'
Learn More

2-Butylfuran, TRC

Product Code. 30138057
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138057 100 mg 100mg
30136349 50 mg 50mg
30010002 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138057 Supplier Toronto Research Chemicals Supplier No. TRCB701653100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4466-24-4
Molecular Formula C8 H12 O
Molecular Weight (g/mol) 124.18
Synonym 2-Butylfuran, 2-n-Butylfuran, Furan, 2-butyl-
IUPAC Name 2-butylfuran
SMILES CCCCc1occc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H12O/c1-2-3-5-8-6-4-7-9-8/h4,6-7H,2-3,5H2,1H3
Formula Weight 124.0888
Chemical Name or Material 2-Butylfuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.