accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N2H2H2HO2H2H2H2H

CAS RN 4472-41-7

IUPAC Name: N,N-di(²H₃)methyl(²H)formamide
Synonyms: deuterated dmf
Molecular Weight (g/mol): 80.14
Molecular Formula: C3H7NO
InChi Key: ZMXDDKWLCZADIW-YYWVXINBSA-N
SMILES: [2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]
Molecular Weight (g/mol) 
  • (7)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (3)
  • (7)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (5)