CAS RN 452-73-3
IUPAC Name:
2-chloro-4-fluoro-1-methylbenzene
Synonyms:
2-chloro-4-fluoro-1-methylbenzene
Molecular Weight (g/mol):
144.57
Molecular Formula:
C7H6ClF
InChi Key:
CSARJIQZOSVYHA-UHFFFAOYSA-N
SMILES:
CC1=CC=C(F)C=C1Cl
1
–
2
of
2
results
1
2-Chloro-4-fluorotoluene, 98%
CAS: 452-73-3 Molecular Formula: C7H6ClF Molecular Weight (g/mol): 144.573 MDL Number: MFCD00000572 InChI Key: CSARJIQZOSVYHA-UHFFFAOYSA-N Synonym: 2-chloro-4-fluorotoluene,benzene, 2-chloro-4-fluoro-1-methyl,4-fluoro-2-chlorotoluene,2-chlor-4-fluor-1-methylbenzol,pubchem1605,2-chloro-4fluorotoluene,acmc-1adyb,4-fluor-2-chlor-toluol,ksc235q0r,csarjiqzosvyha-uhfffaoysa PubChem CID: 96747 IUPAC Name: 2-chloro-4-fluoro-1-methylbenzene SMILES: CC1=C(C=C(C=C1)F)Cl