missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorotoluene, 98%, ACROS Organics™

Product Code. 10033730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10033730 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10033730 Supplier Thermo Scientific Acros Supplier No. 311240250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 452-73-3
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H6ClF
Quantity 25 g
Synonym 2-chloro-4-fluorotoluene, benzene, 2-chloro-4-fluoro-1-methyl, 4-fluoro-2-chlorotoluene, 2-chlor-4-fluor-1-methylbenzol, pubchem1605, 2-chloro-4fluorotoluene, acmc-1adyb, 4-fluor-2-chlor-toluol, ksc235q0r, csarjiqzosvyha-uhfffaoysa
InChI Key CSARJIQZOSVYHA-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)F)Cl
IUPAC Name 2-chloro-4-fluoro-1-methylbenzene
Molecular Weight (g/mol) 144.573
PubChem CID 96747
Formula Weight 144.58
Chemical Name or Material 2-Chloro-4-fluorotoluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.