accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFCH3O

CAS RN 455-91-4

IUPAC Name: 1-(3-fluoro-4-methoxyphenyl)ethan-1-one
Synonyms: 1-(3-fluoro-4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: LQASUDVYVOFKNK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1F)C(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results