accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOBr

CAS RN 4559-96-0

IUPAC Name: 1-(4-bromophenyl)-4-chlorobutan-1-one
Synonyms: 1-(4-bromophenyl)-4-chlorobutan-1-one
Molecular Weight (g/mol): 261.54
Molecular Formula: C10H10BrClO
InChi Key: WJKPUMBLABGUCQ-UHFFFAOYSA-N
SMILES: ClCCCC(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results