accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSO

CAS RN 4565-29-1

IUPAC Name: 5-acetylthiophene-2-carbaldehyde
Synonyms: 5-acetylthiophene-2-carbaldehyde
Molecular Weight (g/mol): 154.18
Molecular Formula: C7H6O2S
InChi Key: ISGDPVVLVWQENT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(S1)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results