accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHNCl

CAS RN 458532-98-4

IUPAC Name: (3-chloropyridin-4-yl)boronic acid
Synonyms: 3-chloropyridin-4-ylboronic acid
Molecular Weight (g/mol): 157.36
Molecular Formula: C5H5BClNO2
InChi Key: JLIHQPWLAOYTRH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=NC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results