missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropyridine-4-boronic acid, 96%, Thermo Scientific™

Product Code. 15469637
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469637 250 mg 250mg
15459637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469637 Supplier Thermo Scientific Alfa Aesar Supplier No. H52864.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458532-98-4
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD03425942
InChI Key JLIHQPWLAOYTRH-UHFFFAOYSA-N
Synonym 3-chloro-4-pyridineboronic acid, 3-chloropyridin-4-yl boronic acid, 3-chloropyridine-4-boronic acid, 3-chloro-4-pyridyl boronic acid, 4-borono-3-chloropyridine, 3-chloropyridin-4-yl-4-boronic acid, boronic acid, 3-chloro-4-pyridinyl, 3-chloropyridine-4-boronic acid hydrate, acmc-209k4r, 3-chloropyridine-4-boronic acid,
PubChem CID 3861877
IUPAC Name (3-chloropyridin-4-yl)boronic acid
SMILES B(C1=C(C=NC=C1)Cl)(O)O

Specifications

Melting Point 95°C to 97°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 157.37
Percent Purity 96%
Chemical Name or Material 3-Chloropyridine-4-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.