accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CS

CAS RN 459409-74-6

IUPAC Name: 4,4,5,5-tetramethyl-2-(5-phenylthiophen-2-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(5-phenylthiophen-2-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 286.2
Molecular Formula: C16H19BO2S
InChi Key: BOTJOCNWKJVWRR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results