accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 460-43-5

CH3OFFF

CAS RN 460-43-5

IUPAC Name: 1,1,1-trifluoro-2-methoxyethane
Synonyms: 1,1,1-trifluoro-2-methoxyethane
Molecular Weight (g/mol): 114.07
Molecular Formula: C3H5F3O
InChi Key: OHLVGBXMHDWRRX-UHFFFAOYSA-N
SMILES: COCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results