accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO(R)H2N

CAS RN 4606-65-9

IUPAC Name: (3R)-3-(hydroxymethyl)piperidin-1-ium
Synonyms: (3R)-3-(hydroxymethyl)piperidin-1-ium
Molecular Weight (g/mol): 116.18
Molecular Formula: C6H14NO
InChi Key: VUNPWIPIOOMCPT-ZCFIWIBFSA-O
SMILES: OC[C@@H]1CCC[NH2+]C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)