accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOH

CAS RN 464-72-2

IUPAC Name: tetraphenylethane-1,2-diol
Synonyms: tetraphenylethane-1,2-diol
Molecular Weight (g/mol): 366.46
Molecular Formula: C26H22O2
InChi Key: MFEWNFVBWPABCX-UHFFFAOYSA-N
SMILES: OC(C1=CC=CC=C1)(C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results