CAS RN 4670-10-4
CAS RN 4670-10-4
IUPAC Name:
2-(3,5-dimethoxyphenyl)acetate
Synonyms:
2-(3,5-dimethoxyphenyl)acetate
Molecular Weight (g/mol):
195.2
Molecular Formula:
C10H11O4
InChi Key:
FFPAFDDLAGTGPQ-UHFFFAOYSA-M
SMILES:
COC1=CC(OC)=CC(CC([O-])=O)=C1