missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenylacetic acid, 98%

Product Code. 11421697
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421697 5 g 5g
11411697 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11421697 Supplier Thermo Scientific Alfa Aesar Supplier No. B25672.06
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 4670-10-4 | C8H14O | 126.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4670-10-4
MDL Number MFCD00016827

Specifications

Melting Point 102°C to 104°C
Molecular Formula C10H11O4
Quantity 5 g
Synonym 3,5-dimethoxyphenylacetic acid, 2-3,5-dimethoxyphenyl acetic acid, 3,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 3,5-dimethoxy, 3,5-dimethoxyphenylaceticacid, 3,5-dimethoxybenzeneacetic acid, 3,5-dimethoxy-phenylacetic acid, 3,5-dimethoxy phenylacetic acid, 3,5-dimethoxy phenyl acetic acid
InChI Key FFPAFDDLAGTGPQ-UHFFFAOYSA-M
SMILES COC1=CC(=CC(=C1)CC(=O)O)OC
IUPAC Name 2-(3,5-dimethoxyphenyl)acetic acid
Molecular Weight (g/mol) 195.20
PubChem CID 138316
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxyphenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.